Learn More
Medchemexpress LLC PROTAC EZH2 Degrader-1 | 2641601-67-2 | 99.4% | 942.15 | 25 MG
SDP
Catalog No. 5000363676
Encompass_Preferred
PROTAC EZH2 Degrader-1 (Compound 150d) is a potent PROTAC EZH2 Degrader that inhibits EZH2 methyltransferase activity with an IC50 of 2.7 nM. EZH2 is critical in many tumorigenesis and development processes.
  • Potently inhibits EZH2 methyltransferase activity (IC50: 2.7 nM).
  • Demonstrates anti-proliferative activity against human EOL1, MV4-11, Pfeiffer, RS4-11, and WSUDLCL2 cells.
  • Inhibits cell viability in H128-LM, H128-BPM, H128 wildtype, and H128-Mut cells.
  • Enhances sensitivity of H128-LM cells to Carboplatin, VP16, and VM26.
  • Exhibits antitumor activity in H128-LM xenograft mouse models, prolonging survival when combined with Carboplatin, VP16, or VM26.
Catalog No. 50-003-63676 Supplier Medchemexpress LLC Supplier No. 25MGHY147525
PROTAC EZH2 Degrader-1 (Compound 150d) is a potent PROTAC EZH2 Degrader that inhibits EZH2 methyltransferase activity with an IC50 of 2.7 nM. EZH2 is critical in many tumorigenesis and development processes.
  • Potently inhibits EZH2 methyltransferase activity (IC50: 2.7 nM).
  • Demonstrates anti-proliferative activity against human EOL1, MV4-11, Pfeiffer, RS4-11, and WSUDLCL2 cells.
  • Inhibits cell viability in H128-LM, H128-BPM, H128 wildtype, and H128-Mut cells.
  • Enhances sensitivity of H128-LM cells to Carboplatin, VP16, and VM26.
  • Exhibits antitumor activity in H128-LM xenograft mouse models, prolonging survival when combined with Carboplatin, VP16, or VM26.