Learn More
Medchemexpress LLC PROTAC Her3 Degrader-8 | 2103331-95-7 | C49H55N11O6S | 5 MG
SDP
Catalog No. 5000376759
Encompass_Preferred
PROTAC Her3 Degrader-8 (Compound PP2) is a Her3 PROTAC degrader designed to promote the ubiquitination and degradation of Her3. This compound is valuable for research into lung adenocarcinoma and ovarian cancer.
  • Promotes the ubiquitination and degradation of Her3
  • Useful in the study of lung adenocarcinoma
  • Useful in the study of ovarian cancer
  • Composed of a Her3 ligand, an E3 ligase ligand, and a linker
  • Degrades Her3 protein in PC9-GR4 or Ovacar 8 cell lines
Catalog No. 50-003-76759 Supplier Medchemexpress LLC Supplier No. HY1537965MG
PROTAC Her3 Degrader-8 (Compound PP2) is a Her3 PROTAC degrader designed to promote the ubiquitination and degradation of Her3. This compound is valuable for research into lung adenocarcinoma and ovarian cancer.
  • Promotes the ubiquitination and degradation of Her3
  • Useful in the study of lung adenocarcinoma
  • Useful in the study of ovarian cancer
  • Composed of a Her3 ligand, an E3 ligase ligand, and a linker
  • Degrades Her3 protein in PC9-GR4 or Ovacar 8 cell lines