Learn More
LifeSensors PROTAC In Vitro Ubiquitination Assay Kit
SDP
Catalog No. NC3737901
Encompass_Preferred
Proteolysis Targeting Chimeras (PROTACs) have emerged as novel therapeutics to target the undruggable proteome. PROTACs are heterobifunctional small molecules that both simultaneously bind to a target protein and a ubiquitin (Ub) E3 ligase. This subsequently leads to ubiquitination and degradation of the target protein. The In vitro ubiquitination kit has been developed to establish a high throughput approach that can accurately predict PROTAC efficiency by monitoring the protein’s intrinsic ability to get ubiquitinated. We offer this kit for three E3 ubiquitin ligases: Cereblon, VHL, and HDM2 to monitor PROTAC mediated ubiquitination for target of choice.For evaluating molecular glues, we recommend using our MG780: Molecular Glue In Vitro Ubiquitination Assay Kit. This kit is specially designed to measure the performance of molecular glues In vitro.
Catalog No. NC3737901 Supplier LifeSensors Supplier No. PA0770P100R
Proteolysis Targeting Chimeras (PROTACs) have emerged as novel therapeutics to target the undruggable proteome. PROTACs are heterobifunctional small molecules that both simultaneously bind to a target protein and a ubiquitin (Ub) E3 ligase. This subsequently leads to ubiquitination and degradation of the target protein. The In vitro ubiquitination kit has been developed to establish a high throughput approach that can accurately predict PROTAC efficiency by monitoring the protein’s intrinsic ability to get ubiquitinated. We offer this kit for three E3 ubiquitin ligases: Cereblon, VHL, and HDM2 to monitor PROTAC mediated ubiquitination for target of choice.For evaluating molecular glues, we recommend using our MG780: Molecular Glue In Vitro Ubiquitination Assay Kit. This kit is specially designed to measure the performance of molecular glues In vitro.