Learn More
Medchemexpress LLC PROTAC IRAK4 degrader-1 | 2360533-90-8 | 99.8% | 50 MG
SDP
Catalog No. 5000338958
Encompass_Preferred
PROTAC IRAK4 degrader-1 (compound I-210) is a Cereblon-based IRAK4 PROTAC. It is composed of an IRAK4 ligand, an E3 ligase ligand, and a PROTAC linker. This product is for research use only.
  • Cereblon-based IRAK4 PROTAC
  • Composed of an IRAK4 ligand, an E3 ligase ligand, and a PROTAC linker
  • Induces IRAK4 protein degradation in OCI-LY-10 cells
Catalog No. 50-003-38958 Supplier Medchemexpress LLC Supplier No. HY12996650MG
PROTAC IRAK4 degrader-1 (compound I-210) is a Cereblon-based IRAK4 PROTAC. It is composed of an IRAK4 ligand, an E3 ligase ligand, and a PROTAC linker. This product is for research use only.
  • Cereblon-based IRAK4 PROTAC
  • Composed of an IRAK4 ligand, an E3 ligase ligand, and a PROTAC linker
  • Induces IRAK4 protein degradation in OCI-LY-10 cells