Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PROTAC JAK1 degrader 1 | 3052553-20-2 | 100.0% | 791.85 | 5 MG
SDP

Catalog No. 5000392741
Encompass_Preferred

PROTAC JAK1 degrader 1 (compound 10c) is an effective and selective PROTAC JAK1 degrader with a DC50 of 214 nM. It also exhibits antitumor activity. The product is soluble in DMSO and can be formulated for in vivo applications. Proper storage includes protection from light and nitrogen atmosphere for solid and solvent forms.

  • Effective and selective JAK1 degrader
  • DC50 of 214 nM
  • Exhibits antitumor activity
  • Soluble in DMSO
  • Suitable for in vivo applications
  • Recommended storage conditions protect product integrity

Catalog No. 50-003-92741 Supplier Medchemexpress LLC Supplier No. HY1626085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC JAK1 degrader 1 (compound 10c) is an effective and selective PROTAC JAK1 degrader with a DC50 of 214 nM. It also exhibits antitumor activity. The product is soluble in DMSO and can be formulated for in vivo applications. Proper storage includes protection from light and nitrogen atmosphere for solid and solvent forms.

  • Effective and selective JAK1 degrader
  • DC50 of 214 nM
  • Exhibits antitumor activity
  • Soluble in DMSO
  • Suitable for in vivo applications
  • Recommended storage conditions protect product integrity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.