Learn More
Medchemexpress LLC PROTAC KRAS G12C degrader-1 | 2984236-79-3 | 99.15% | 917.47 | 50 MG
SDP
Catalog No. 5000352044
Encompass_Preferred
PROTAC KRAS G12C degrader-1 is a Cereblon-based KRASG12C PROTAC degrader. It induces CRBN/KRASG12C dimerization and degrades GFP-KRASG12C in reporter cells. This compound is for research use only.
  • Engages CRBN in cells
  • Binds KRASG12C in vitro
  • Induces CRBN/KRASG12C dimerization
  • Degrades GFP-KRASG12C in reporter cells in a CRBN-dependent manner
Catalog No. 50-003-52044 Supplier Medchemexpress LLC Supplier No. HY13918650MG
Add to Cart
Add to Cart
PROTAC KRAS G12C degrader-1 is a Cereblon-based KRASG12C PROTAC degrader. It induces CRBN/KRASG12C dimerization and degrades GFP-KRASG12C in reporter cells. This compound is for research use only.
  • Engages CRBN in cells
  • Binds KRASG12C in vitro
  • Induces CRBN/KRASG12C dimerization
  • Degrades GFP-KRASG12C in reporter cells in a CRBN-dependent manner