Learn More
Medchemexpress LLC PROTAC KRAS G12C degrader-1 | 2984236-79-3 | 99.2% | C50H54ClFN8O6 | 10MG
SDP
Catalog No. 5000261838
Encompass_Preferred
PROTAC KRAS G12C degrader-1 is a cereblon-recruiting PROTAC that induces ubiquitin-proteasome degradation of KRAS G12C mutant proteins in cellular assays. It promotes CRBN-KRASG12C ternary complex formation and is used to evaluate degradation potency and downstream signaling in reporter and cancer cell lines.
  • Cereblon-based recruitment mechanism for targeted degradation.
  • Effective degradation of GFP-KRASG12C in reporter cells.
  • High purity suitable for biochemical and cellular experiments.
  • Stable as a powder under recommended cold storage conditions.
  • Available in multiple small-scale quantities for research applications.
Catalog No. 50-002-61838 Supplier Medchemexpress LLC Supplier No. HY13918610MG
PROTAC KRAS G12C degrader-1 is a cereblon-recruiting PROTAC that induces ubiquitin-proteasome degradation of KRAS G12C mutant proteins in cellular assays. It promotes CRBN-KRASG12C ternary complex formation and is used to evaluate degradation potency and downstream signaling in reporter and cancer cell lines.
  • Cereblon-based recruitment mechanism for targeted degradation.
  • Effective degradation of GFP-KRASG12C in reporter cells.
  • High purity suitable for biochemical and cellular experiments.
  • Stable as a powder under recommended cold storage conditions.
  • Available in multiple small-scale quantities for research applications.