Learn More
Medchemexpress LLC PROTAC MDM2 Degrader-1 | 2249944-98-5 | 98.1% | 1 MG
SDP
Catalog No. 5000338822
Encompass_Preferred
PROTAC MDM2 Degrader-1 (Compound 15a) is an MDM2 PROTAC degrader. The structures of both linker ends are MDM2 ligands. This compound can block the binding of p53-MDM2 and also degrade the target MDM2 protein by utilizing the E3 ligase function of MDM2 itself, thereby exhibiting an anti-tumor effect. It is intended for research use only.
  • MDM2 PROTAC degrader
  • Blocks the binding of p53-MDM2
  • Degrades the target MDM2 protein
  • Exhibits anti-tumor effect
  • Intended for research use only
Catalog No. 50-003-38822 Supplier Medchemexpress LLC Supplier No. HY1288401MG
PROTAC MDM2 Degrader-1 (Compound 15a) is an MDM2 PROTAC degrader. The structures of both linker ends are MDM2 ligands. This compound can block the binding of p53-MDM2 and also degrade the target MDM2 protein by utilizing the E3 ligase function of MDM2 itself, thereby exhibiting an anti-tumor effect. It is intended for research use only.
  • MDM2 PROTAC degrader
  • Blocks the binding of p53-MDM2
  • Degrades the target MDM2 protein
  • Exhibits anti-tumor effect
  • Intended for research use only