Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PROTAC MDM2 Degrader-1 | 2249944-98-5 | 98.1% | 10 MG
SDP

Catalog No. 5000338208
Encompass_Preferred

PROTAC MDM2 Degrader-1 (Compound 15a) is an MDM2 PROTAC degrader. It blocks p53-MDM2 binding and degrades the target MDM2 protein by utilizing the E3 ligase function of MDM2 itself, thereby exerting an anti-tumor effect.

  • Targets MDM2
  • Inhibits MDM2 expression and promotes p53 expression in A549 cells
  • Molecular weight: 1463.33
  • Formula: C74H84Cl4N10O13
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO at 100 mg/mL (68.34 mM)
  • Soluble in 10% DMSO and 90% Corn Oil (≥ 0.62 mg/mL)

Catalog No. 50-003-38208 Supplier Medchemexpress LLC Supplier No. HY12884010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC MDM2 Degrader-1 (Compound 15a) is an MDM2 PROTAC degrader. It blocks p53-MDM2 binding and degrades the target MDM2 protein by utilizing the E3 ligase function of MDM2 itself, thereby exerting an anti-tumor effect.

  • Targets MDM2
  • Inhibits MDM2 expression and promotes p53 expression in A549 cells
  • Molecular weight: 1463.33
  • Formula: C74H84Cl4N10O13
  • Appearance: Solid
  • Color: White to off-white
  • Soluble in DMSO at 100 mg/mL (68.34 mM)
  • Soluble in 10% DMSO and 90% Corn Oil (≥ 0.62 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.