Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protac MDM2 degrader-1 | 2249944-98-5 | MFCD32197288 | 98.1% | 1463.33 | C74H84Cl4N10O13 | 5 MG
SDP

Catalog No. 5000264026
Encompass_Preferred

PROTAC MDM2 Degrader-1 is a research-grade PROTAC designed to target MDM2 for induced degradation and p53 modulation in cell-based studies. It is supplied as a high-purity solid in small milligram quantities for preclinical research.

  • Small-molecule PROTAC that induces MDM2 degradation for research use.
  • High purity: 98.1%.
  • Molecular weight: 1463.33 Da; formula: C74H84Cl4N10O13.
  • Appearance: solid, supplied in milligram quantities.
  • Storage: store under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Available package sizes: 1 mg, 5 mg, 10 mg, 25 mg; larger sizes by quote.

Catalog No. 50-002-64026 Supplier Medchemexpress LLC Supplier No. HY1288405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC MDM2 Degrader-1 is a research-grade PROTAC designed to target MDM2 for induced degradation and p53 modulation in cell-based studies. It is supplied as a high-purity solid in small milligram quantities for preclinical research.

  • Small-molecule PROTAC that induces MDM2 degradation for research use.
  • High purity: 98.1%.
  • Molecular weight: 1463.33 Da; formula: C74H84Cl4N10O13.
  • Appearance: solid, supplied in milligram quantities.
  • Storage: store under nitrogen; in solvent -80°C (6 months) or -20°C (1 month).
  • Available package sizes: 1 mg, 5 mg, 10 mg, 25 mg; larger sizes by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.