Learn More
Medchemexpress LLC PROTAC MDM2 Degrader-2 | 2249944-99-6 | 95.0% | 10 MG
SDP
Catalog No. 5000338172
Encompass_Preferred
PROTAC MDM2 Degrader-2 is an MDM2 PROTAC degrader that induces the self-ubiquitination and degradation of MDM2, leading to the upregulation of p53 protein. It exhibits anti-tumor activity and is used in cancer research.
  • Induces self-ubiquitination and degradation of MDM2
  • Upregulates p53 protein
  • Exhibits anti-tumor activity
  • Used in cancer research
  • Inhibits MDM2 protein in A549 cells
  • Increases p53 in A549 cells
Catalog No. 50-003-38172 Supplier Medchemexpress LLC Supplier No. HY12884110MG
PROTAC MDM2 Degrader-2 is an MDM2 PROTAC degrader that induces the self-ubiquitination and degradation of MDM2, leading to the upregulation of p53 protein. It exhibits anti-tumor activity and is used in cancer research.
  • Induces self-ubiquitination and degradation of MDM2
  • Upregulates p53 protein
  • Exhibits anti-tumor activity
  • Used in cancer research
  • Inhibits MDM2 protein in A549 cells
  • Increases p53 in A549 cells