Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PROTAC MDM2 Degrader-3 | 2249750-23-8 | 95.0% | 1 MG
SDP

Catalog No. 5000339914
Encompass_Preferred

PROTAC MDM2 Degrader-3 is an MDM2 degrader based on PROTAC technology. It is composed of a potent MDM2 inhibitor, a linker, and an MDM2 ligand for E3 ubiquitin ligase. This compound is intended for research use only.

  • Utilizes PROTAC technology
  • Composed of a potent MDM2 inhibitor, linker, and MDM2 ligand for E3 ubiquitin ligase
  • Molecular weight of 1437.25
  • Chemical formula: C72H78Cl4N8O15
  • Appearance: white to off-white solid
  • Target: MDM2

Catalog No. 50-003-39914 Supplier Medchemexpress LLC Supplier No. HY1288421MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PROTAC MDM2 Degrader-3 is an MDM2 degrader based on PROTAC technology. It is composed of a potent MDM2 inhibitor, a linker, and an MDM2 ligand for E3 ubiquitin ligase. This compound is intended for research use only.

  • Utilizes PROTAC technology
  • Composed of a potent MDM2 inhibitor, linker, and MDM2 ligand for E3 ubiquitin ligase
  • Molecular weight of 1437.25
  • Chemical formula: C72H78Cl4N8O15
  • Appearance: white to off-white solid
  • Target: MDM2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.