Learn More
Medchemexpress LLC HY-141881 5mg , PROTAC-O4I2 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1418815MG
Encompass_Preferred
HY-141881 5mg PROTAC-O4I2 CAS: PROTAC-O4I2 is a PROTAC targets splicing factor 3B1 (SF3B1). PROTAC-O4I2 induces FLAG-SF3B1 degradation with an IC50 value of 0.244 μM in K562 cells. PROTAC-O4I2 also induces cellular apoptosis in K562 WT cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1843