Learn More
Medchemexpress LLC PROTAC RAR Degrader-1 | 1351169-27-1 | 97.9% | 917.14 | 10 MG
SDP
Catalog No. 5000439004
Encompass_Preferred
PROTAC RAR Degrader-1 (Compound 9) is a potent and selective RAR PROTAC Degrader that incorporates apoptotic protein inhibitors (IAPs) ligands, also known as SNIPERs. It reduces RARα levels in HT1080 cells in a concentration-dependent manner, a process that is blocked by the proteasome inhibitor MG132. This product is suitable for studying nuclear receptor-related diseases.
  • Potent and selective RAR PROTAC Degrader.
  • Consists of apoptotic protein inhibitors (IAPs) ligands (SNIPERs).
  • Reduces RARα levels in HT1080 cells in a concentration-dependent manner.
  • Useful in the study of nuclear receptor-related diseases.
Catalog No. 50-004-39004 Supplier Medchemexpress LLC Supplier No. HY11184410MG
Add to Cart
Add to Cart
PROTAC RAR Degrader-1 (Compound 9) is a potent and selective RAR PROTAC Degrader that incorporates apoptotic protein inhibitors (IAPs) ligands, also known as SNIPERs. It reduces RARα levels in HT1080 cells in a concentration-dependent manner, a process that is blocked by the proteasome inhibitor MG132. This product is suitable for studying nuclear receptor-related diseases.
  • Potent and selective RAR PROTAC Degrader.
  • Consists of apoptotic protein inhibitors (IAPs) ligands (SNIPERs).
  • Reduces RARα levels in HT1080 cells in a concentration-dependent manner.
  • Useful in the study of nuclear receptor-related diseases.