Learn More
Medchemexpress LLC PROTAC RIPK DEGRADER-2
SDP
Catalog No. NC1855132
Encompass_Preferred
A nonpeptidic PROTAC which potently targets serine-threonine kinase RIPK2 and has highly selective for RIPK2 degradation.
Catalog No. NC1855132 Supplier Medchemexpress LLC Supplier No. HY11186610MG
A nonpeptidic PROTAC which potently targets serine-threonine kinase RIPK2 and has highly selective for RIPK2 degradation.