Learn More
Medchemexpress LLC HY-103636 10mg Medchemexpress, PROTAC Sirt2 Degrader-1 CAS:2098487-75-1 Purity:>98%
SDP
Catalog No. 501968146
Encompass_Preferred
Medchemexpress, HY-103636 10mg PROTAC Sirt2 Degrader-1 CAS:2098487-75-1 PROTAC Sirt2 Degrader-1 is a SirReal-based PROTAC, acts as a Sirt2 degrader, composed of a highly potent and isotype-selective Sirt2 inhibitor, a linker, and a bona fide cereblon ligand for E3 ubiquitin ligase. PROTAC Sirt2 Degrader-1 shows an IC50 of 0.25 μM for Sirt2, with no effect on Sirt1/Sirt3 (IC50s > 100 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8146 Supplier Medchemexpress LLC Supplier No. HY10363610MG
Add to Cart
Add to Cart
Medchemexpress, HY-103636 10mg PROTAC Sirt2 Degrader-1 CAS:2098487-75-1 PROTAC Sirt2 Degrader-1 is a SirReal-based PROTAC, acts as a Sirt2 degrader, composed of a highly potent and isotype-selective Sirt2 inhibitor, a linker, and a bona fide cereblon ligand for E3 ubiquitin ligase. PROTAC Sirt2 Degrader-1 shows an IC50 of 0.25 μM for Sirt2, with no effect on Sirt1/Sirt3 (IC50s > 100 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.