Learn More
Medchemexpress LLC PROTAC SOS1 degrader-1 TFA | 98.4% | C59H77ClF4N10O6S | 1 MG
SDP
Catalog No. 5000355925
Encompass_Preferred
PROTAC SOS1 degrader-1 TFA is a potent PROTAC SOS1 degrader with a DC50 of 98.4 nM. It demonstrates antiproliferation activity in cancer cells with various KRAS mutations and shows an antitumor effect with low toxicity.
  • Potent SOS1 degrader
  • Effective in cancer cells with KRAS mutations
  • Exhibits low toxicity
  • Solid, white to off-white appearance
Catalog No. 50-003-55925 Supplier Medchemexpress LLC Supplier No. HY145737A1MG
PROTAC SOS1 degrader-1 TFA is a potent PROTAC SOS1 degrader with a DC50 of 98.4 nM. It demonstrates antiproliferation activity in cancer cells with various KRAS mutations and shows an antitumor effect with low toxicity.
  • Potent SOS1 degrader
  • Effective in cancer cells with KRAS mutations
  • Exhibits low toxicity
  • Solid, white to off-white appearance