Learn More
Medchemexpress LLC PROTAC STING Degrader-1 | 2762552-74-7 | C34H33N7O10 | 5 MG
SDP
Catalog No. 5000375304
Encompass_Preferred
PROTAC STING Degrader-1 is a PROTAC degrader targeting the STING pathway, demonstrating high anti-inflammatory efficacy with a DC50 of 3.2 μM. It is valuable for studying diseases such as acute kidney injury and inflammatory bowel diseases.
  • Targets the STING pathway with a DC50 of 3.2 μM.
  • Exhibits high anti-inflammatory efficacy.
  • Useful for studying acute kidney injury and inflammatory bowel diseases.
  • Downregulates STING pathway signals (IFN-β, IL-6, CXCL10) in THP-1 cells.
  • Degrades STING dose- and time-dependently.
  • Demonstrates no significant cytotoxicity below 10 μM.
  • Alleviates ischemic symptoms and reduces kidney weights in acute kidney injury mouse models.
  • Ameliorates severity of DSS-induced colitis in mice.
Catalog No. 50-003-75304 Supplier Medchemexpress LLC Supplier No. HY1506085MG
PROTAC STING Degrader-1 is a PROTAC degrader targeting the STING pathway, demonstrating high anti-inflammatory efficacy with a DC50 of 3.2 μM. It is valuable for studying diseases such as acute kidney injury and inflammatory bowel diseases.
  • Targets the STING pathway with a DC50 of 3.2 μM.
  • Exhibits high anti-inflammatory efficacy.
  • Useful for studying acute kidney injury and inflammatory bowel diseases.
  • Downregulates STING pathway signals (IFN-β, IL-6, CXCL10) in THP-1 cells.
  • Degrades STING dose- and time-dependently.
  • Demonstrates no significant cytotoxicity below 10 μM.
  • Alleviates ischemic symptoms and reduces kidney weights in acute kidney injury mouse models.
  • Ameliorates severity of DSS-induced colitis in mice.