Learn More
Medchemexpress LLC PROTAC STING degrader-3 | 3109967-69-0 | 98.0% | 752.73 | 1 MG
SDP
Catalog No. 5000385348
Encompass_Preferred
PROTAC STING degrader-3 is a STING PROTAC degrader that induces STING degradation via the ubiquitin-proteoteasome pathway. It exerts anti-inflammatory effects by inhibiting STING/TBK1/NF-κB signaling and has renal protective effects, making it useful in the study of acute kidney injury (AKI).
  • STING ligand
  • Cereblon ligand
  • Linker
Catalog No. 50-003-85348 Supplier Medchemexpress LLC Supplier No. HY1734141MG
PROTAC STING degrader-3 is a STING PROTAC degrader that induces STING degradation via the ubiquitin-proteoteasome pathway. It exerts anti-inflammatory effects by inhibiting STING/TBK1/NF-κB signaling and has renal protective effects, making it useful in the study of acute kidney injury (AKI).
  • STING ligand
  • Cereblon ligand
  • Linker