Learn More
Medchemexpress LLC Protac α-synuclein degrader 3 | 2412273-77-7 | ≥98.0% | C51H68N6O11S2 | 10MG
SDP
Catalog No. 5000259210
Encompass_Preferred
PROTAC α-synuclein degrader 3 is a proteolysis-targeting chimera developed for research applications to induce ubiquitination and proteasomal degradation of α-synuclein, useful in Parkinson's disease model studies. It has documented in vitro and in vivo formulation solubility and recommended storage conditions for stable handling.
  • Selective α-synuclein degrader that promotes ubiquitination and proteasomal degradation.
  • Demonstrated to reduce α-synuclein levels (≈65% reduction at 1 μM).
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Formulatable for in vivo dosing in multiple vehicle systems at ≥2.5 mg/mL.
  • Stable when stored as powder at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for clinical or therapeutic use.
Catalog No. 50-002-59210 Supplier Medchemexpress LLC Supplier No. HY13600110MG
PROTAC α-synuclein degrader 3 is a proteolysis-targeting chimera developed for research applications to induce ubiquitination and proteasomal degradation of α-synuclein, useful in Parkinson's disease model studies. It has documented in vitro and in vivo formulation solubility and recommended storage conditions for stable handling.
  • Selective α-synuclein degrader that promotes ubiquitination and proteasomal degradation.
  • Demonstrated to reduce α-synuclein levels (≈65% reduction at 1 μM).
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Formulatable for in vivo dosing in multiple vehicle systems at ≥2.5 mg/mL.
  • Stable when stored as powder at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not intended for clinical or therapeutic use.