Learn More
Medchemexpress LLC Protac α-synuclein degrader 3 | 2412273-77-7 | 99.6% | 1005.25 g·mol⁻¹ | C51H68N6O11S2 | 5 MG
SDP
Catalog No. 5000221862
Encompass_Preferred
PROTAC α-synuclein degrader 3 is a small-molecule proteolysis targeting chimera (PROTAC) designed to promote ubiquitination and proteasomal degradation of α-synuclein aggregates. It is intended for research applications in Parkinson's disease and related neurodegeneration studies, provided as a solid with high chemical characterization for reproducible experimental use.
  • Promotes ubiquitination and degradation of α-synuclein.
  • High chemical purity (~99.6%).
  • Suitable for cell-based and biochemical assays.
  • Available in multiple package sizes for experimental flexibility.
  • Characterized by formula C51H68N6O11S2 and molecular weight 1005.25 g·mol⁻¹.
Catalog No. 50-002-21862 Supplier Medchemexpress LLC Supplier No. HY1360015MG
PROTAC α-synuclein degrader 3 is a small-molecule proteolysis targeting chimera (PROTAC) designed to promote ubiquitination and proteasomal degradation of α-synuclein aggregates. It is intended for research applications in Parkinson's disease and related neurodegeneration studies, provided as a solid with high chemical characterization for reproducible experimental use.
  • Promotes ubiquitination and degradation of α-synuclein.
  • High chemical purity (~99.6%).
  • Suitable for cell-based and biochemical assays.
  • Available in multiple package sizes for experimental flexibility.
  • Characterized by formula C51H68N6O11S2 and molecular weight 1005.25 g·mol⁻¹.