Learn More
Medchemexpress LLC HY-107911 100mg , Protamine sulfate CAS:9009-65-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107911100MG
Encompass_Preferred
HY-107911 100mg Protamine sulfate CAS: 9009-65-8 Protamine sulfate, polycationic peptide and a antiheparin agent, could neutralize the anticoagulant action of heparin and enhances lipid-mediated gene transfer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1270