Learn More
Sigma Aldrich Fine Chemicals Biosciences Protamine sulfate, MFCD00132091, 10g
Catalog No. NC1592299
Encompass
Meets USP testing specifications, Synonym: Protamine sulfate, Salmine sulfate from salmon sperm; Protamine sulfate is a cationic polypeptide having a molecular weight of 5 kDa, isolated from salmon sperm.
Catalog No. NC1592299 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P336910G
Meets USP testing specifications, Synonym: Protamine sulfate, Salmine sulfate from salmon sperm; Protamine sulfate is a cationic polypeptide having a molecular weight of 5 kDa, isolated from salmon sperm.