Learn More
Medchemexpress LLC Protamine sulfate 200mg | 9009-65-8 | 200MG
SDP
Catalog No. 5000242847
Encompass_Preferred
Protamine sulfate polycationic peptide and a antiheparin agent could neutralize the anticoagulant action of heparin and enhances lipid-mediated gene transfer[1 [2 [3
Catalog No. 50-002-42847 Supplier Medchemexpress LLC Supplier No. HY107911200MG
Protamine sulfate polycationic peptide and a antiheparin agent could neutralize the anticoagulant action of heparin and enhances lipid-mediated gene transfer[1 [2 [3