Learn More
Medchemexpress LLC Protamine sulfate | 9009-65-8 | 100.0% | 5 kDa (≈5,000 g/mol) | No single chemical formula - mixture of peptides (~5 kDa) | 1 G
SDP
Catalog No. 5000242286
Encompass_Preferred
Protamine sulfate is a polycationic peptide (≈5 kDa) research reagent used as an antiheparin agent to neutralize the anticoagulant effects of unfractionated heparin and to enhance lipid-mediated gene transfer. It is supplied as a mixture of peptides derived from salmon sperm and is intended for research use only.
  • Neutralizes heparin anticoagulant activity.
  • Enhances lipid-mediated gene transfer.
  • Soluble in water and PBS at ~33.33 mg/mL; may require ultrasonication.
  • Powder stable at -20°C for up to 3 years; solutions stable at -80°C for 6 months or -20°C for 1 month.
  • Approximate molecular weight ~5 kDa (polypeptide mixture).
Catalog No. 50-002-42286 Supplier Medchemexpress LLC Supplier No. HY1079111G
Protamine sulfate is a polycationic peptide (≈5 kDa) research reagent used as an antiheparin agent to neutralize the anticoagulant effects of unfractionated heparin and to enhance lipid-mediated gene transfer. It is supplied as a mixture of peptides derived from salmon sperm and is intended for research use only.
  • Neutralizes heparin anticoagulant activity.
  • Enhances lipid-mediated gene transfer.
  • Soluble in water and PBS at ~33.33 mg/mL; may require ultrasonication.
  • Powder stable at -20°C for up to 3 years; solutions stable at -80°C for 6 months or -20°C for 1 month.
  • Approximate molecular weight ~5 kDa (polypeptide mixture).