Learn More
Medchemexpress LLC Protamine sulfate | 9009-65-8 | MFCD00132091 | 100.0% | 5,000-10,000 Da | 500 MG
SDP
Catalog No. 5000242227
Encompass_Preferred
Protamine sulfate is a polycationic peptide used as an antiheparin agent that neutralizes the anticoagulant activity of heparin and can enhance lipid-mediated gene transfer. Supplied as a purified research reagent, its molecular weight and composition are polydisperse and batch dependent; consult the certificate of analysis for exact properties.
  • Neutralizes heparin anticoagulation
  • Enhances lipid-mediated gene transfer
  • High-purity peptide suitable for research applications
  • Water-soluble and easy to prepare in aqueous buffers
  • Polydisperse molecular weight; use batch certificate of analysis for exact values
  • Widely used in heparin reversal and nucleic acid delivery studies
Catalog No. 50-002-42227 Supplier Medchemexpress LLC Supplier No. HY107911500MG
Protamine sulfate is a polycationic peptide used as an antiheparin agent that neutralizes the anticoagulant activity of heparin and can enhance lipid-mediated gene transfer. Supplied as a purified research reagent, its molecular weight and composition are polydisperse and batch dependent; consult the certificate of analysis for exact properties.
  • Neutralizes heparin anticoagulation
  • Enhances lipid-mediated gene transfer
  • High-purity peptide suitable for research applications
  • Water-soluble and easy to prepare in aqueous buffers
  • Polydisperse molecular weight; use batch certificate of analysis for exact values
  • Widely used in heparin reversal and nucleic acid delivery studies