Learn More
Sigma Aldrich Fine Chemicals Biosciences Protamine sulfate salt from salmon, 53597-25-4, MFCD00132091, 5 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P40205G
Encompass
Grade X, amorphous powder, Synonym: Salmine from salmon, Salmine. May be used for the removal of DNA from protein samples or for the purification of DNA binding proteins. It may be used as an alternative to polybrene in retroviral-mediated gene transfer.
Catalog No. NC0480516