Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protamine sulfate

Catalog No. AAJ6292609
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6292609 10 g
AAJ6292603 1 g
AAJ6292606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6292609 Supplier Thermo Scientific Chemicals Supplier No. J6292609
Add to Cart

Reverses the anticoagulant effects of heparin | CAS: 9009-65-8

It is suitable for personal care, cosmetic and pharmaceutical applications. It is used widely to reverse the anticoagulant effects of heparin. It acts as a heparin antagonist. It is also a weak anticoagulant. It may be used for the removal of DNA from protein samples or the purification of DNA binding proteins. Protamine sulfate may also be used as an alternative to polybrene in retroviral mediated gene transfer. Protamine sulfate has antibacterial properties and inhibits pepsin activity. Protamine sulfate and other polycations have been shown to inhibit the classical pathway of complement. It neutralizes the anticoagulant activity of heparin, and inhibits turnover of lipoproteins by lipoprotein lipase. The aggregation of erythrocytes in the presence of protamine sulfate, for application to the prediction of MN phenotypes, has been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is sparingly soluble in water.

Notes
Hygroscopic. Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9009-65-8
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132091

Specifications

Color White
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information It is sparingly soluble in water.
Physical Form Powder
Chemical Name or Material Protamine sulfate
RTECSNumber UK9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.