Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protamine Sulfate, USP/NF Grade, MP Biomedicals™

Catalog No. ICN10275205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10275205 5 g
ICN10275225 25 g
ICN10275280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10275205 Supplier MP Biomedicals, Inc Supplier No. 0210275205
Add to Cart
Add to Cart
Molecular biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9009-65-8
Molecular Weight (g/mol) 0.00
Synonym Angiociclan, Dilangio

Specifications

Color White
Physical Form Solid
Quantity 5 g
Grade NF/USP
Chemical Name or Material Protamine Sulfate
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.