Learn More
Grainger ProtCase 4.30 in PwrClwLtcSys/PdLk Or
Catalog No. 502766559
Encompass_Preferred
The Grainger 16Z503 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a plastic organizer case for storage and portability. The sockets are made from chrome vanadium steel for durability and have a matte finish to prevent corrosion. This socket set is ideal for automotive and light industrial applications.
Catalog No. 50-276-6559 Supplier Grainger Supplier No. 16Z503
The Grainger 16Z503 is a Westward brand 1/4-inch drive socket set. It contains both SAE and metric sockets ranging from 4mm to 15mm and 1/8 inch to 9/16 inch. The set has a total of 38 pieces and comes in a plastic organizer case for storage and portability. The sockets are made from chrome vanadium steel for durability and have a matte finish to prevent corrosion. This socket set is ideal for automotive and light industrial applications.