Learn More
Med Vet International Instrument Protector 3M Comply 6-2/3 inch Length, 13913, 1000/Case
Catalog No. 502830505
Encompass_Preferred
Can be used for both steam and EO sterilization Holds the instrument in an open position for proper sterilization For use inside a single peel pouch
Catalog No. 50-283-0505 Supplier Med Vet International Supplier No. 1112461CS
Can be used for both steam and EO sterilization Holds the instrument in an open position for proper sterilization For use inside a single peel pouch