Learn More
Bio Rad Laboratories PROTEAN II XI COMB
Catalog No. 509012242
Encompass
Comb; Bio-Rad; Protean II xi; 15-well; 1.5mm
Catalog No. 50-901-2242 Supplier Bio Rad Laboratories Supplier No. 1651873
Comb; Bio-Rad; Protean II xi; 15-well; 1.5mm