Learn More
OriGene ProteanFect TuffCell Mouse Immunocyte Transfection Kit - 0.02 mL
SDP
Catalog No. 5000600189
Encompass
A non-viral, non-electroporation, non-liposomal transfection kit using engineered mammalian proteins, tailored for high-efficiency delivery in mouse primary immune cellsComponents: 4 vials- Reagent A, 1.2 mL- Reagent B, 0.02 mL- Reagent C, 0.3 mL- EGFP mRNA, 5 µLStability: 12 months (-20 + 5°C)Storage: The Entire Kit can be stored at -80°C.Acceptable storage:Reagent A (2-8°C); Reagent B (-20°C); Reagent C (2-8°C); EGFP mRNA (-80°C)
Catalog No. 50-006-00189 Supplier OriGene Supplier No. TT300008S
A non-viral, non-electroporation, non-liposomal transfection kit using engineered mammalian proteins, tailored for high-efficiency delivery in mouse primary immune cellsComponents: 4 vials- Reagent A, 1.2 mL- Reagent B, 0.02 mL- Reagent C, 0.3 mL- EGFP mRNA, 5 µLStability: 12 months (-20 + 5°C)Storage: The Entire Kit can be stored at -80°C.Acceptable storage:Reagent A (2-8°C); Reagent B (-20°C); Reagent C (2-8°C); EGFP mRNA (-80°C)