Learn More
OriGene ProteanFect TuffCell Transfection Kit - 0.02 mL
SDP
Catalog No. 5000600188
Encompass_Preferred
A non-viral, non-electroporation, non-liposomal transfection kit using engineered mammalian proteins, tailored for high-efficiency delivery into a wide range of primary cells and difficult to transfect cell lines such as immune cells, stem cells, and neuronal cells.Components: 5 vials- Reagent A, 1.2 mL- Reagent B, 0.02 mL- Reagent C, 0.5 mL- EGFP mRNA, 5 µL- EGFP pDNA, 8 µLStability 12 months (-20 + 5°C)Storage The Entire Kit can be stored at -80°C.Acceptable storage:Reagent A (2-8°C); Reagent B (-20°C); Reagent C (2-8°C); EGFP mRNA (-80°C); EGFP pDNA (-20°C)
Catalog No. 50-006-00188 Supplier OriGene Supplier No. TT300007S
A non-viral, non-electroporation, non-liposomal transfection kit using engineered mammalian proteins, tailored for high-efficiency delivery into a wide range of primary cells and difficult to transfect cell lines such as immune cells, stem cells, and neuronal cells.Components: 5 vials- Reagent A, 1.2 mL- Reagent B, 0.02 mL- Reagent C, 0.5 mL- EGFP mRNA, 5 µL- EGFP pDNA, 8 µLStability 12 months (-20 + 5°C)Storage The Entire Kit can be stored at -80°C.Acceptable storage:Reagent A (2-8°C); Reagent B (-20°C); Reagent C (2-8°C); EGFP mRNA (-80°C); EGFP pDNA (-20°C)