Learn More
Enzo Life Sciences Proteasome 19S Rpn2/S1 subunit (human) monoclonal antibody (112-1) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW92700100
Encompass_Preferred
Clone: 112-1. Isotype: IgG1. Immunogen: Recombinant full-length human Rpn2 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes the Rpn2/S1 subunit of the 19S regulator complex. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1676