Learn More
Enzo Life Sciences Proteasome 19S Rpn12/S14 subunit (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW88150025
Encompass_Preferred
Immunogen: Recombinant full length proteasome 26S (19S regulator complex) (Rpn12/S14 subunit). Species reactivity: Human. Specificity: Recognizes Rpn12/S14 subunit of the 19S regulator complex. Applications: IHC, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1640