Learn More
Enzo Life Sciences Proteasome 19S Rpt1/S7 subunit monoclonal antibody (MSS1-104) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW88250100
Encompass_Preferred
Clone: MSS1-104. Isotype: IgG1. Immunogen: Recombinant human Rpt1 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Specificity: Recognizes the Rpt1/S7 subunit of the 19S regulator complex. Applications: WB. Purity: Purified. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1642