Learn More
Enzo Life Sciences Proteasome 19S Rpn12/S14 subunit (human) monoclonal antibody (P31-27) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW88350025
Encompass_Preferred
Clone: p31-27. Isotype: IgG1. Immunogen: Recombinant Rpn12 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human. Specificity: Recognizes human proteasome 26S (19S regulator complex) (Rpn12/S14 subunit). Applications: IHC, WB. Purity: Partially purified. Formulation: Liquid. Containing 10mM sodium azide.
Catalog No. 50-201-1645