Learn More
Enzo Life Sciences Proteasome 19S ATPase subunit Rpt6 monoclonal antibody (p45-110) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW92650025
Encompass_Preferred
Clone: p45-110. Isotype: IgG2b. Immunogen: Recombinant human Rpt6 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat. Specificity: Recognizes the Rpt6/S subunit of the 19S regulator complex. Applications: IHC, IP, WB. Purity: Partially purified. Formulation: Liquid. In PBS, pH 7.4, containing 10mM sodium azide.
Catalog No. 50-201-1673