Learn More
Enzo Life Sciences Proteasome 19S Rpn10/S5a subunit monoclonal antibody (S5a-18) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW92500100
Encompass_Preferred
Clone: S5a-18. Isotype: IgG1. Immunogen: Recombinant full-length human Rpn10 protein. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Rat, Bovine. Specificity: Recognizes the Rpn10/S5a subunit of the 19S regulator complex. Applications: IP, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1671