Learn More
Enzo Life Sciences Proteasome 19S Rpt5/S6a subunit monoclonal antibody (TBP1-19) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW87700025
Encompass_Preferred
Clone: TBP1-19. Isotype: IgG2b. Immunogen: Recombinant full-length human Rpt5. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Rabbit. Specificity: Recognizes the Rpt5/S6a subunit of the 19S regulatory subunit. Applications: IHC, WB. Purity: Partially purified ascites. Formulation: Liquid. Contains 10mM sodium azide.
Catalog No. 50-201-1638