Learn More
Enzo Life Sciences Proteasome 19S Rpt3/S6b subunit monoclonal antibody (TBP7-27) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW87650025
Encompass_Preferred
Clone: TBP7-27. Isotype: IgG1. Immunogen: Recombinant full-length human Rpt3. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human Rabbit. Specificity: Recognizes the Rpt3/S6b subunit of the 19S regulator complex. Applications: IF, IHC, IP, WB. Purity: Partially purified. Formulation: Liquid. Contains 10mM sodium azide.
Catalog No. 50-201-1636