Learn More
Enzo Life Sciences Proteasome 19S Rpn12/S14 subunit polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLPW04400025
Encompass_Preferred
Immunogen: Full-length Arabidopsis proteasome 26S (19S regulator complex) (Rpn12a/S14 subunit). Species reactivity: Arabidopsis thaliana. Specificity: Recognizes the Rpn12a/S14 subunit of the 19S regulator complex. Applications: WB. Formulation: Liquid. Serum.
Catalog No. 50-201-1524