Learn More
Enzo Life Sciences Proteasome 19S Rpn8/S12 subunit (human) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81800025
Encompass_Preferred
Immunogen: Recombinant human proteasome 26S (19S regulator complex) (Rpn8/S12 subunit) (aa 1-205). Species reactivity: Human. Specificity: Recognizes Rpn8/S12 subunit of the 19S regulator complex. Applications: IHC, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1588