Learn More
Enzo Life Sciences Proteasome 19S Rpt2/S4 subunit (human) polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences BMLPW05300025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 368-383 of human proteasome 19S (Rpt2/S4 subunit). Source: Purified from rabbit serum. Species reactivity: Human. Applications: WB. Purity: Partially purified by salt precipitation. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1535