Learn More
Enzo Life Sciences Proteasome 19S Rpt3/S6b subunit polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82500100
Encompass_Preferred
Immunogen: Recombinant protein corresponding to the N-terminal 100 amino acids of the YTA2 protein. Species reactivity: Human Yeast. Specificity: Recognizes the Rpt3/S6b subunit of the 19S regulator complex. Applications: IHC, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1608