Learn More
Enzo Life Sciences Proteasome 19S Rpt5/S6a subunit polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82450100
Encompass_Preferred
Immunogen: Recombinant protein corresponding to the N-terminal 100 amino acids of Yta1. Species reactivity: Yeast. Specificity: Recognizes the Rpt5/S6a subunit of the 19S regulator complex. Applications: WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1606