Learn More
Enzo Life Sciences Proteasome 19S Rpn7/S10a subunit polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82250100
Encompass_Preferred
Immunogen: Recombinant full length human proteasome 26S (19S regulator complex) (Rpn7/S10a subunit). Species reactivity: Human Yeast. Specificity: Recognizes the Rpt7/S10a subunit of the 19S regulator complex. Applications: IP, WB. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1598