Learn More
Enzo Life Sciences Proteasome 19S Rpt1/S7 subunit (yeast) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW82550100
Encompass_Preferred
Immunogen: Recombinant protein corresponding to the N-terminal 100 amino acids of YTA3. Species reactivity: Yeast. Specificity: Recognizes the Rpt1/S7 subunit of the 19S regulator complex. Applications: WB. Formulation: Liquid. Neat serum.
Catalog No. 50-201-1610