Learn More
Enzo Life Sciences Proteasome 20S alpha subunit (human) monoclonal antibody (HP810) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW82650100
Encompass_Preferred
Clone: HP810. Isotype: IgG. Immunogen: Human erythrocyte-derived proteasomes. Species reactivity: Human. Specificity: Recognizes the α subunit of the 20S proteasome. Applications: ICC, IF, IP. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-201-1612